Stampa…

Anti-SNX6 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIS1014-97C2FITCEA 852 EUR
USBIS1014-97C2FITC
Anti-SNX6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: SNX6 (sorting nexin 6)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR