Stampa…

Anti-SNX6 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS1014-97C2EA 587 EUR
USBIS1014-97C2
Anti-SNX6 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: SNX6 (sorting nexin 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR