Anti-IgG F(ab) (H+L) Rabbit Polyclonal Antibody (Biotin)
Catalog # USBII1903-18X
Supplier: US Biological
- Antibody type:Secondary
- Antigen symbol:IgG F(ab) (H+L)
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Chicken
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, 10mg/ml BSA, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
About this item
Recommended dilution: ELISA: 1:16000-1:80000
Type: Secondary
Antigen: IgG Fab H+L
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken