Stampa…
Anti-Endophilin 1, Mid, SH3 Domain Rabbit polyclonal antibody
Fornitore: United States Biological
USBIE2291-48DEA 592 EUR
USBIE2291-48D
Anti-Endophilin 1, Mid, SH3 Domain Rabbit polyclonal antibody
Anticorpi
Recommended dilution: ELISA: 0. 01-1 ?g/ml; Western Blot: 1-3?g/ml
Type: Primary
Antigen: Endophilin 1, Mid, SH3 Domain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat