Stampa…

Anti-ENDOG Rabbit polyclonal antibody

Fornitore: United States Biological
USBIE2291-40FEA 450 EUR
USBIE2291-40F
Anti-ENDOG Rabbit polyclonal antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: ENDOG (endonuclease G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR