Stampa…

Anti-EMCN Mouse monoclonal antibody [clone: 10C74]

USBIE2291-34BEA 0 EUR
USBIE2291-34B
Anti-EMCN Mouse monoclonal antibody [clone: 10C74]
Type: Primary
Antigen: EMCN (endomucin)
Clonality: monoclonal
Clone: 10C74
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR