Stampa…

Anti-Endomorphin 1 Rabbit polyclonal antibody

Fornitore: United States Biological
USBIE2291-04EA 979 EUR
USBIE2291-04
Anti-Endomorphin 1 Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: Endomorphin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR