Stampa…

Anti-ENG Rabbit polyclonal antibody

Fornitore: United States Biological
USBIE2287-03CEA 675 EUR
USBIE2287-03C
Anti-ENG Rabbit polyclonal antibody
Anticorpi
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: ENG (endoglin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR