Stampa…

Anti-Endogenous Retrovirus Type K, Human, gag Mouse Monoclonal Antibody [clone: 5i76]

Fornitore: United States Biological
USBIE2286-19EA 719 EUR
USBIE2286-19
Anti-Endogenous Retrovirus Type K, Human, gag Mouse Monoclonal Antibody [clone: 5i76]
Anticorpi
Recommended dilution: ELISA: 1:2000; Western Blot: 1:8000

Type: Primary
Antigen: Endogenous Retrovirus Type K, Human, gag
Clonality: Monoclonal
Clone: 5i76
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR