Stampa…

Anti-ERVW-1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: United States Biological
USBIE2286-05A-HRPEA 852 EUR
USBIE2286-05A-HRP
Anti-ERVW-1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: ERVW-1 (endogenous retrovirus group W, member 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR