Stampa…

Anti-ERVW-1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIE2286-05A-FITCEA 852 EUR
USBIE2286-05A-FITC
Anti-ERVW-1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: ERVW-1 (endogenous retrovirus group W, member 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR