Stampa…

Anti-OPN3 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIE2285-15EA 675 EUR
USBIE2285-15
Anti-OPN3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: OPN3 (opsin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR