Stampa…
Anti-Encephalitis, St. Louis Mouse monoclonal antibody [clone: 10D421]
Fornitore: United States Biological
USBIE2280-50AEA 776 EUR
USBIE2280-50A
Anti-Encephalitis, St. Louis Mouse monoclonal antibody [clone: 10D421]
Anticorpi
Type: Primary
Antigen: Encephalitis, St. Louis
Clonality: monoclonal
Clone: 10D421
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Visualizza i codici