Anti-COL3 Rabbit Polyclonal Antibody
Catalog # USBIC7510-44E
Supplier: US Biological
- Antibody type:Primary
- Antigen name:collagen, type III
- Antigen symbol:COL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Form:liquid
- Storage temperature:Cool
- Concentration:~4,5 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
About this item
Recommended dilution: Immunochemistry: 1:250-1:1000 ELISA: 1:100-1:400; Immunofluorescence: 1:80; Radio Immunoassay: 1:25-1:100
Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat