Anti-ALDH7A1 Rabbit Polyclonal Antibody
USBI123218
Specifications
- Antibody type:Primary
- Antigen name:aldehyde dehydrogenase 7 family, member A1
- Antigen symbol:ALDH7A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 511
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ALDH7A1 (aldehyde dehydrogenase 7 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human