Stampa…
Anti-Spike Mouse monoclonal antibody
MEDMMM-0151-PEA 329 EUR
MEDMMM-0151-P
Anti-Spike Mouse monoclonal antibody
Type: Primary
Antigen: Spike
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici