Stampa…
Anti-Cleaved Tubulin Rabbit polyclonal antibody unconjugated
Fornitore: Medimabs
MEDMMM-0143EA 339 EUR
MEDMMM-0143
Anti-Cleaved Tubulin Rabbit polyclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: Cleaved Tubulin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human