Stampa…
Anti-Cleaved Tau Rabbit polyclonal antibody unconjugated
Fornitore: Medimabs
MEDMMM-0141EA 280 EUR
MEDMMM-0141
Anti-Cleaved Tau Rabbit polyclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: Cleaved Tau
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human