Stampa…
Anti-SMAP2 Rabbit Polyclonal Antibody
Fornitore: Medimabs
MEDMMM-0138EA 243 EUR
MEDMMM-0138
Anti-SMAP2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: SMAP2 (small ArfGAP2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human