Stampa…

Anti-ARF6 Rabbit Polyclonal Antibody

Fornitore: Medimabs
MEDMMM-0126EA 243 EUR
MEDMMM-0126
Anti-ARF6 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ARF6 (ADP-ribosylation factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR