Stampa…
Anti-Chicken Dscam Mouse Monoclonal Antibody
Fornitore: Medimabs
MEDMMM-0114-PEA 326 EUR
MEDMMM-0114-P
Anti-Chicken Dscam Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Chicken Dscam
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici