Stampa…
Anti-Phosphorylated Prion Protein Rabbit Polyclonal Antibody
Fornitore: Medimabs
MEDMMM-0112EA 290 EUR
MEDMMM-0112
Anti-Phosphorylated Prion Protein Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Phosphorylated Prion Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity:
Visualizza i codici