Stampa…
Anti-HHV6 1E2 protein Mouse Monoclonal Antibody
Fornitore: Medimabs
MEDMMM-0071-PEA 329 EUR
MEDMMM-0071-P
Anti-HHV6 1E2 protein Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: HHV6 1E2 protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici