Stampa…
Anti-HACE1 Mouse Monoclonal Antibody
Fornitore: Medimabs
MEDMMM-0066-PEA 292 EUR
MEDMMM-0066-P
Anti-HACE1 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: HACE1 (HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human