Stampa…

Anti-TOH Mouse Monoclonal Antibody

Fornitore: Medimabs
MEDMMM-0063EA 345 EUR
MEDMMM-0063
Anti-TOH Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: TOH
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR