Stampa…

Anti-PSEN1 Mouse Monoclonal Antibody

Fornitore: Medimabs
MEDMMM-0021EA 281 EUR
MEDMMM-0021
Anti-PSEN1 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR