Stampa…
Anti-RPTPs Mouse Monoclonal Antibody
Fornitore: Medimabs
MEDMMM-0020-PEA 509 EUR
MEDMMM-0020-P
Anti-RPTPs Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: RPTPs
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici