Stampa…
Anti-APP Mouse monoclonal antibody unconjugated
Fornitore: Medimabs
MEDMMM-0015-PEA 474 EUR
MEDMMM-0015-P
Anti-APP Mouse monoclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse