Stampa…
Anti-DBH Mouse monoclonal antibody unconjugated
Fornitore: Medimabs
MEDMMM-0008-PEA 394 EUR
MEDMMM-0008-P
Anti-DBH Mouse monoclonal antibody unconjugated
Anticorpi
Type: Primary
Antigen: DBH (dopamine beta-hydroxylase (dopamine beta-monooxygenase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat