Stampa…
Anti-Substance P Mouse Monoclonal Antibody
Fornitore: Medimabs
MEDMMM-0001-1EA 402 EUR
MEDMMM-0001-1
Anti-Substance P Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: Substance P
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Visualizza i codici