Stampa…
Anti-SFT2D3 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBI041637EA 587 EUR
USBI041637
Anti-SFT2D3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Type: Primary
Antigen: SFT2D3 (SFT2 domain containing 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human