Stampa…
Anti-SFSWAP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornitore: United States Biological
USBI041635-APEA 852 EUR
USBI041635-AP
Anti-SFSWAP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SFSWAP (splicing factor, suppressor of white-apricot homolog (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human