Stampa…

Anti-SRSF3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: United States Biological
USBI041633-PEEA 852 EUR
USBI041633-PE
Anti-SRSF3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SRSF3 (serine/arginine-rich splicing factor 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR