Stampa…
Anti-SERTAD2 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBI041593EA 587 EUR
USBI041593
Anti-SERTAD2 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERTAD2 (SERTA domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human