Stampa…
Anti-SERPINI2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: United States Biological
USBI041592-PEEA 852 EUR
USBI041592-PE
Anti-SERPINI2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINI2 (serpin peptidase inhibitor, clade I (pancpin), member 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human