Stampa…
Anti-SERPINI2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornitore: United States Biological
USBI041592-HRPEA 852 EUR
USBI041592-HRP
Anti-SERPINI2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERPINI2 (serpin peptidase inhibitor, clade I (pancpin), member 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human