Stampa…
Anti-SERPINE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornitore: United States Biological
USBI041585-FITCEA 852 EUR
USBI041585-FITC
Anti-SERPINE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SERPINE1 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human