Stampa…
Anti-SERPINB9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: United States Biological
USBI041583-PEEA 852 EUR
USBI041583-PE
Anti-SERPINB9 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Do not freeze!
Type: Primary
Antigen: SERPINB9 (serpin peptidase inhibitor, clade B (ovalbumin), member 9)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human