Stampa…
Anti-SERPINB8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornitore: United States Biological
USBI041582-APCEA 852 EUR
USBI041582-APC
Anti-SERPINB8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINB8 (serpin peptidase inhibitor, clade B (ovalbumin), member 8)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human