Stampa…
Anti-SERPINB8 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBI041582EA 587 EUR
USBI041582
Anti-SERPINB8 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERPINB8 (serpin peptidase inhibitor, clade B (ovalbumin), member 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human