Stampa…

Anti-SERPINA5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: United States Biological
USBI041575-FITCEA 852 EUR
USBI041575-FITC
Anti-SERPINA5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SERPINA5 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR