Stampa…
Anti-SERINC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornitore: United States Biological
USBI041568-HRPEA 852 EUR
USBI041568-HRP
Anti-SERINC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERINC1 (serine incorporator 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human