Stampa…
Anti-SEPT8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornitore: United States Biological
USBI041565-FITCEA 852 EUR
USBI041565-FITC
Anti-SEPT8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPT8 (septin 8)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human