Stampa…
Anti-SEPT6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornitore: United States Biological
USBI041564-APCEA 852 EUR
USBI041564-APC
Anti-SEPT6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPT6 (septin 6)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse