Stampa…
Anti-SEPT5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: United States Biological
USBI041563-PEEA 852 EUR
USBI041563-PE
Anti-SEPT5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SEPT5 (septin 5)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human