Stampa…
Anti-SEPT5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornitore: United States Biological
USBI041563-APCEA 852 EUR
USBI041563-APC
Anti-SEPT5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SEPT5 (septin 5)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human