Stampa…
Anti-SEPT2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornitore: United States Biological
USBI041561-APEA 852 EUR
USBI041561-AP
Anti-SEPT2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPT2 (septin 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human