Stampa…
Anti-SEPT10 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBI041558-BIOTINEA 852 EUR
USBI041558-BIOTIN
Anti-SEPT10 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SEPT10 (septin 10)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human