Stampa…
Anti-SEPT1 Rabbit Polyclonal Antibody (Biotin)
Fornitore: United States Biological
USBI041557-BIOTINEA 852 EUR
USBI041557-BIOTIN
Anti-SEPT1 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SEPT1 (septin 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human