Stampa…
Anti-SEPP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornitore: United States Biological
USBI041555-PEEA 852 EUR
USBI041555-PE
Anti-SEPP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPP1 (selenoprotein P, plasma, 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human